siRNA Details
Gene Symbol: Lypd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029806.1 | NP_084082.1 | 5 | 616 | ||
GenBank mRNA | AK008654.1 | - | 5 | 619 | ||
BC107186.2 | - | 5 | 616 | 40050796 | ||
BC107187.2 | - | 5 | 616 | 40050800 | ||
BC119485.1 | - | 5 | 616 | 40050803 | ||
BC119486.1 | - | 5 | 616 | 40050804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
