siRNA Details
Gene Symbol: 2610028H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029816.2 | NP_084092.2 | 5 | 617 | ||
GenBank mRNA | AK011591.1 | - | 5 | 573 | ||
AK161552.1 | - | 5 | 427 | |||
BC051454.1 | - | 5 | 554 | 6308081 | ||
BQ714071.1 | - | 5 | 636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
