siRNA Details
Gene Symbol: 2700049A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163378.1 | NP_001156850.1 | 19 | 3677 | ||
NM_029818.1 | NP_084094.1 | 19 | 3677 | |||
GenBank mRNA | AK012399.1 | - | 4 | 357 | ||
AK172976.1 | - | 19 | 3327 | |||
BC043104.1 | - | 19 | 3677 | |||
BC150744.1 | - | 19 | 3045 | 9087655 |

More Information
Other Species Targeted

Gene Ontology Categories:
