siRNA Details
Gene Symbol: 2700049A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163378.1 | NP_001156850.1 | 15, 16 | 3248 | ||
NM_029818.1 | NP_084094.1 | 15, 16 | 3248 | |||
GenBank mRNA | AK172976.1 | - | 15, 16 | 2898 | ||
BC043104.1 | - | 15, 16 | 3248 | |||
BC150744.1 | - | 15, 16 | 2616 | 9087655 |
