siRNA Details
Gene Symbol: Urah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029821.2 | NP_084097.1 | 3 | 424 | ||
GenBank mRNA | AK004470.1 | - | 3 | 642 | ||
AK013117.1 | - | 3 | 426 | |||
BC051545.1 | - | 3 | 592 | 5255010 | ||
DQ225768.1 | - | 3 | 299 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
