siRNA Details
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039201.1 | NP_001034290.1 | 4 | 480 | ||
NM_001039202.1 | NP_001034291.1 | 4 | 441 | |||
NM_029826.2 | NP_084102.1 | 4 | 441 | |||
GenBank mRNA | AK002837.1 | - | 4 | 429 | ||
AK014208.1 | - | 4 | 435 | |||
AK137285.1 | - | 4 | 433 | |||
AK147794.1 | - | 462 | ||||
AK151285.1 | - | 4 | 429 | |||
AK152162.1 | - | 4 | 429 | |||
AK152259.1 | - | 4 | 429 | |||
BC058177.1 | - | 4 | 415 | 5250739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
