siRNA Details
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029832.2 | NP_084108.1 | 5 | 688 | ||
GenBank mRNA | BC051541.1 | - | 3 | 277 | ||
BC115616.2 | - | 5 | 688 | 40102754 | ||
BC115617.1 | - | 5 | 688 | 40102757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
