siRNA Details
Gene Symbol: Slc9a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB089793.1 | - | 9 | 760 | ||
AF482993.1 | - | 8 | 724 | |||
AK049410.1 | - | 9 | 756 | |||
AK051791.1 | - | 8 | 675 | |||
AK143281.1 | - | 9 | 729 | |||
AK160826.1 | - | 7 | 598 | |||
AK173069.1 | - | 9 | 1096 | |||
AK216559.1 | - | 2 | 128 | |||
BC030879.1 | - | 9 | 759 | 4241988 | ||
BC034508.1 | - | 2 | 160 | |||
BC058947.1 | - | 9 | 730 | 5702781 |

More Information
Other Species Targeted

