siRNA Details
Gene Symbol: Tstd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029840.1 | NP_084116.1 | 2 | 220 | ||
GenBank mRNA | AI287097.1 | - | 2 | 220 | ||
AK011611.1 | - | 2 | 135 | |||
AK154853.1 | - | 2 | 207 | |||
AK218953.1 | - | 1 | 60 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
