siRNA Details
Gene Symbol: Mrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029844.3 | NP_084120.1 | 1 | 302 | ||
GenBank mRNA | AK003912.2 | - | 1 | 303 | ||
AK158113.1 | - | 1 | 176 | |||
AY079153.1 | - | 1 | 79 | |||
AY079154.1 | - | 1 | 62 | |||
BC027543.1 | - | 1 | 156 | 1477097 |

More Information
Other Species Targeted

Gene Ontology Categories:
