siRNA Details
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166024.1 | NP_001159496.1 | 13, 14 | 1454 | ||
NM_023854.2 | NP_076343.2 | 12, 13 | 1412 | |||
GenBank mRNA | AF229439.1 | - | 12, 13 | 1221 | ||
AK009565.1 | - | 13, 14 | 1278 | |||
AK149837.1 | - | 12, 13 | 1247 | |||
AK168489.1 | - | 12, 13 | 1222 | |||
AK169144.1 | - | 12, 13 | 1235 | |||
BC005495.1 | - | 12, 13 | 1260 | 3487811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
