siRNA Details
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101.3 | NP_001092.1 | 6 | 1430 | ||
GenBank mRNA | AK025375.1 | - | 6 | 1429 | ||
AK058019.1 | - | 6 | 1430 | |||
AK098751.1 | - | 7 | 1233 | |||
AK130062.1 | - | 6 | 1432 | |||
AK130157.1 | - | 6 | 1431 | |||
AK222925.1 | - | 6 | 1433 | |||
AK223032.1 | - | 6 | 1428 | |||
AK223055.1 | - | 6 | 1428 | |||
AK225414.1 | - | 6 | 1265 | |||
AK304552.1 | - | 7 | 1359 | |||
AK308277.1 | - | 6 | 1399 | |||
AK309997.1 | - | 7 | 1472 | |||
BC001301.1 | - | 6 | 1413 | 3451917 | ||
BC002409.2 | - | 6 | 1418 | 2961617 | ||
BC004251.1 | - | 6 | 1407 | 3623067 | ||
BC008633.1 | - | 5 | 1327 | |||
BC009636.1 | - | 1 | 164 | |||
BC012854.1 | - | 5 | 1301 | |||
BC013380.2 | - | 6 | 1422 | 4025526 | ||
BC013835.1 | - | 6 | 1419 | |||
BC014861.1 | - | 6 | 1420 | 3870487 | ||
BC016045.1 | - | 6 | 1421 | 4564532 | ||
V00478.1 | - | 3 | 593 | |||
X00351.1 | - | 6 | 1387 | |||
X63432.1 | - | 6 | 1418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
