siRNA Details
Gene Symbol: Cep83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029852.2 | NP_084128.2 | 2 | 439 | ||
GenBank mRNA | AK011135.1 | - | 2 | 511 | ||
AK015007.1 | - | 2 | 576 | |||
AK141627.1 | - | 2 | 560 | |||
BC028960.1 | - | 1 | 140 | |||
BC043468.1 | - | 1 | 53 | |||
BC070464.1 | - | 2 | 439 | 30608497 |
