siRNA Details
Gene Symbol: Ocel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293800.1 | NP_001280729.1 | 1 | 669 | ||
NM_029865.2 | NP_084141.2 | 1 | 669 | |||
GenBank mRNA | AK020528.1 | - | 1 | 383 | ||
AK143433.1 | - | 622 | ||||
AK155219.1 | - | 1 | 629 | |||
AK163762.1 | - | 670 | ||||
BB766991.1 | - | 1 | 239 | |||
BC019407.1 | - | 1 | 625 | 5151818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
