siRNA Details
Gene Symbol: Tmem200a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029881.3 | NP_084157.2 | 2 | 2210 | ||
GenBank mRNA | AK032569.1 | - | 2 | 1810 | ||
AK048673.1 | - | 3 | 1605 | |||
AK141454.1 | - | 2182 | ||||
AK165025.1 | - | 2 | 1378 | |||
AK173299.1 | - | 2 | 2210 | |||
BC141266.1 | - | 2 | 1435 | 9055878 | ||
BC141267.1 | - | 2 | 1435 | 9055879 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
