siRNA Details
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029896.1 | NP_084172.1 | 6 | 865 | ||
GenBank mRNA | AK035109.1 | - | 6 | 867 | ||
AK037620.1 | - | 6 | 872 | |||
AK149692.1 | - | 6 | 866 | |||
AK159240.1 | - | 6 | 865 | |||
BC019115.1 | - | 5 | 391 | |||
BC031502.1 | - | 6 | 555 | |||
BC094419.1 | - | 6 | 828 | |||
BC157943.1 | - | 788 | 9007278 | |||
BC157945.1 | - | 789 | 9007280 | |||
BC158054.1 | - | 789 | 9007356 | |||
EF011615.1 | - | 6 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
