siRNA Details
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130062.1 | - | 6 | 1482 | ||
AK222925.1 | - | 6 | 1483 | |||
AK223055.1 | - | 6 | 1478 | |||
AK225414.1 | - | 6 | 1315 | |||
AK304552.1 | - | 7 | 1409 | |||
AK308277.1 | - | 6 | 1449 | |||
BC002409.2 | - | 6 | 1468 | 2961617 | ||
BC008633.1 | - | 5 | 1377 | |||
BC013835.1 | - | 6 | 1469 | |||
BC016045.1 | - | 6 | 1471 | 4564532 | ||
V00478.1 | - | 3 | 643 | |||
X00351.1 | - | 6 | 1437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
