siRNA Details
Gene Symbol: 9530002B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023865.3 | NP_076354.1 | 4 | 466 | ||
GenBank mRNA | AF319955.1 | - | 4 | 451 | ||
AK020534.1 | - | 4 | 416 | |||
AY077734.1 | - | 4 | 445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
