siRNA Details
Gene Symbol: Jrkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033181.1 | NP_001028353.1 | 1 | 1604 | ||
GenBank mRNA | BC138472.1 | - | 1 | 1604 | 8861492 | |
BC138475.1 | - | 1 | 1604 | 8861495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
