siRNA Details
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029929.3 | NP_084205.3 | 6 | 792 | ||
GenBank mRNA | AF439858.1 | - | 6 | 776 | ||
AK019463.1 | - | 6 | 776 | |||
AK041982.1 | - | 6 | 788 | |||
AK140553.1 | - | 6 | 781 | |||
AK150282.1 | - | 6 | 784 | |||
AK150550.1 | - | 6 | 784 | |||
AK203866.1 | - | 1 | 25 | |||
BC046417.1 | - | 6 | 783 | 4501207 |
