siRNA Details
Gene Symbol: Spns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029932.3 | NP_084208.1 | 1, 2 | 252 | ||
GenBank mRNA | AK020683.1 | - | 1, 2 | 253 | ||
AK156479.1 | - | 1, 2 | 516 | |||
AK156781.1 | - | 1, 2 | 245 | |||
BC150778.1 | - | 1, 2 | 235 | 9087689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
