siRNA Details
Gene Symbol: Bcl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029933.4 | NP_084209.3 | 9 | 3651 | ||
GenBank mRNA | AK084676.1 | - | 2 | 1578 | ||
AK147659.1 | - | 9 | 3651 | |||
AY296061.1 | - | 6 | 3167 | |||
BC019641.1 | - | 1 | 98 | |||
BC137850.1 | - | 6 | 3139 | 8860869 |
