siRNA Details
Gene Symbol: H2afv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029938.1 | NP_084214.1 | 3, 4 | 300 | ||
GenBank mRNA | AK021267.1 | - | 3, 4 | 223 | ||
AK155105.1 | - | 3, 4 | 303 | |||
AK168110.1 | - | 3, 4 | 272 | |||
AK178770.1 | - | 1, 2 | 103 | |||
BC147293.1 | - | 2, 3 | 259 | 9056353 | ||
BC147294.1 | - | 3, 4 | 285 | 9056344 | ||
BC147741.1 | - | 3, 4 | 285 | 9056690 | ||
BC147751.1 | - | 3, 4 | 285 | 9056700 | ||
CJ069840.1 | - | 3, 4 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
