siRNA Details
Gene Symbol: Prss36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081374.1 | NP_001074843.2 | 15 | 2693 | ||
GenBank mRNA | AK021185.1 | - | 5 | 649 | ||
BC044738.2 | - | 5 | 689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
