siRNA Details
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164609.1 | NP_001158081.1 | 17 | 1781 | ||
NM_001164610.1 | NP_001158082.1 | 16 | 1742 | |||
NM_001164611.1 | NP_001158083.1 | 16 | 1739 | |||
NM_029945.3 | NP_084221.2 | 17 | 1829 | |||
GenBank mRNA | AK019466.2 | - | 17 | 1830 | ||
AK031029.1 | - | 16 | 1733 | |||
AK031685.1 | - | 16 | 1741 | |||
AK035892.1 | - | 16 | 1734 | |||
AK036055.1 | - | 16 | 1734 | |||
AK036380.1 | - | 16 | 1738 | |||
AK129354.1 | - | 17 | 1806 | |||
AK138720.1 | - | 16 | 1716 | |||
AK164234.1 | - | 17 | 1773 | |||
AK164456.1 | - | 14 | 2207 | |||
AK183804.1 | - | 2 | 212 | |||
BC026767.1 | - | 16 | 1715 | 5137008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
