siRNA Details
Gene Symbol: Snapc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029949.3 | NP_084225.1 | 4 | 534 | ||
GenBank mRNA | AK007563.1 | - | 4 | 534 | ||
AK010932.1 | - | 4 | 535 | |||
AK016168.1 | - | 4 | 549 | |||
AK019863.2 | - | 4 | 530 | |||
AK031870.1 | - | 4 | 536 | |||
AK031924.1 | - | 3 | 452 | |||
AK049933.1 | - | 4 | 555 | |||
BC048713.1 | - | 4 | 534 | 6330194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
