siRNA Details
Gene Symbol: Arhgef38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029953.1 | NP_084229.1 | 1 | 244 | ||
GenBank mRNA | AK020284.1 | - | 1 | 305 | ||
BC049886.1 | - | 1 | 244 | 5387778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
