siRNA Details
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 8 | 1348 | ||
NM_001109686.2 | NP_001103156.1 | 8 | 1327 | |||
NM_001109687.2 | NP_001103157.1 | 8 | 1348 | |||
NM_172545.4 | NP_766133.2 | 8 | 1327 | |||
GenBank mRNA | AB205007.1 | - | 8 | 1354 | ||
AK049454.1 | - | 5 | 598 | |||
AK052174.1 | - | 8 | 1311 | |||
AK054555.1 | - | 8 | 1296 | |||
AK082062.1 | - | 8 | 1338 | |||
BC056938.1 | - | 6 | 1049 | |||
BC089302.1 | - | 6 | 1195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
