siRNA Details
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 24, 25 | 3483 | ||
NM_001109686.2 | NP_001103156.1 | 23, 24 | 3324 | |||
NM_001109687.2 | NP_001103157.1 | 23, 24 | 3339 | |||
NM_172545.4 | NP_766133.2 | 24, 25 | 3462 | |||
GenBank mRNA | AB205007.1 | - | 24, 25 | 3489 | ||
AK049454.1 | - | 21, 22 | 2733 | |||
AK052174.1 | - | 24, 25 | 3446 | |||
AK082062.1 | - | 24, 25 | 3473 | |||
AK129461.1 | - | 5, 6 | 574 | |||
AK149125.1 | - | 4, 5 | 406 | |||
BC023904.1 | - | 6, 7 | 728 | |||
BC056938.1 | - | 21, 22 | 3040 | |||
BC089302.1 | - | 21, 22 | 3192 |

More Information
Other Species Targeted

Gene Ontology Categories:
