siRNA Details
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 11 | 1748 | ||
NM_001109686.2 | NP_001103156.1 | 10 | 1589 | |||
NM_172545.4 | NP_766133.2 | 11 | 1727 | |||
GenBank mRNA | AB205007.1 | - | 11 | 1754 | ||
AK049454.1 | - | 8 | 998 | |||
AK052174.1 | - | 11 | 1711 | |||
AK054555.1 | - | 11 | 1696 | |||
AK082062.1 | - | 11 | 1738 | |||
BC089302.1 | - | 8 | 1457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
