siRNA Details
Gene Symbol: Lcn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029959.2 | NP_084235.1 | 5 | 464 | ||
GenBank mRNA | AF435738.1 | - | 5 | 464 | ||
AK020305.1 | - | 5 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
