siRNA Details
Gene Symbol: 6030408B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020048.1 | - | 1 | 395 | ||
AK134453.1 | - | 1 | 391 | |||
AK144445.1 | - | 1 | 410 | |||
BC147793.1 | - | 1 | 390 | |||
BC147798.1 | - | 1 | 390 | |||
BC150861.1 | - | 1 | 390 |
