siRNA Details
Gene Symbol: 6030408B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020048.1 | - | 2 | 509 | ||
AK134453.1 | - | 2 | 505 | |||
AK144445.1 | - | 2 | 524 | |||
BC150861.1 | - | 2 | 504 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6030408B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020048.1 | - | 2 | 509 | ||
AK134453.1 | - | 2 | 505 | |||
AK144445.1 | - | 2 | 524 | |||
BC150861.1 | - | 2 | 504 |