siRNA Details
Gene Symbol: 6030468B19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029964.1 | NP_084240.1 | 2, 3 | 186 | ||
GenBank mRNA | AK020081.1 | - | 2, 3 | 111 | ||
BB843270.1 | - | 2, 3 | 187 |

More Information
Other Species Targeted

Gene Ontology Categories:
