siRNA Details
Gene Symbol: Adamtsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029967.3 | NP_084243.3 | 28 | 5342 | ||
GenBank mRNA | AK031246.1 | - | 11 | 1945 | ||
AK132420.1 | - | 16 | 3782 | |||
BC116295.1 | - | 11 | 1936 | |||
BC116296.1 | - | 11 | 1936 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
