siRNA Details
Gene Symbol: Elp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023876.3 | NP_076365.2 | 8, 9 | 1051 | ||
GenBank mRNA | AJ276004.1 | - | 8, 9 | 1050 | ||
AK034040.1 | - | 8, 9 | 1056 | |||
AK034529.1 | - | 8, 9 | 1057 | |||
AK037104.1 | - | 8, 9 | 1075 | |||
AK039447.1 | - | 8, 9 | 1063 | |||
AK039758.1 | - | 8, 9 | 1052 | |||
AK079069.1 | - | 8, 9 | 1066 | |||
AK168964.1 | - | 8, 9 | 1115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
