siRNA Details
Gene Symbol: Elp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023876.3 | NP_076365.2 | 5 | 545 | ||
GenBank mRNA | AJ276004.1 | - | 5 | 544 | ||
AK020742.1 | - | 5 | 553 | |||
AK034040.1 | - | 5 | 550 | |||
AK034529.1 | - | 5 | 551 | |||
AK037104.1 | - | 5 | 569 | |||
AK039447.1 | - | 5 | 557 | |||
AK039758.1 | - | 5 | 546 | |||
AK079069.1 | - | 5 | 560 | |||
AK168964.1 | - | 5 | 609 | |||
EF410086.1 | - | 409 |

More Information
Other Species Targeted

Gene Ontology Categories:
