siRNA Details
Gene Symbol: Mlana | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029993.1 | NP_084269.1 | 3 | 183 | ||
GenBank mRNA | AK020928.1 | - | 3 | 202 | ||
BC111114.1 | - | 3 | 183 | 40053172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
