siRNA Details
Gene Symbol: Krtap19-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130857.2 | NP_570927.1 | 1 | 414 | ||
GenBank mRNA | AF345295.1 | - | 1 | 366 | ||
AK020697.1 | - | 1 | 415 | |||
BC115545.2 | - | 1 | 366 | 40101652 | ||
BC115546.2 | - | 1 | 366 | 40101653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
