siRNA Details
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207633.2 | NP_997516.1 | 2, 3 | 641 | ||
GenBank mRNA | AK020780.1 | - | 2, 3 | 215 | ||
AK030473.1 | - | 2, 3 | 643 | |||
AK045453.1 | - | 2, 3 | 353 | |||
AK053562.1 | - | 2, 3 | 642 | |||
AK079683.1 | - | 2, 3 | 253 | |||
AK143152.1 | - | 2, 3 | 211 | |||
AK160251.1 | - | 3, 4 | 269 | |||
BC048712.1 | - | 2, 3 | 276 | 6328657 |
