siRNA Details
Gene Symbol: Tmem50b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030018.3 | NP_084294.1 | 4 | 400 | ||
GenBank mRNA | AK020977.1 | - | 4 | 405 | ||
AK098154.1 | - | 4 | 364 | |||
AK133966.1 | - | 4 | 428 | |||
AK150773.1 | - | 388 | ||||
AK151796.1 | - | 4 | 430 | |||
AK152190.1 | - | 4 | 416 | |||
AK152767.1 | - | 4 | 416 | |||
AK155082.1 | - | 4 | 453 | |||
AK170684.1 | - | 4 | 395 | |||
AK172335.1 | - | 4 | 388 | |||
BC004841.1 | - | 4 | 403 | 2649264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
