siRNA Details
Gene Symbol: Cutal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021330.1 | - | 2 | 211 | ||
AK033810.1 | - | 2 | 289 | |||
AK085616.1 | - | 2 | 250 | |||
BC021771.1 | - | 2 | 167 | 4194825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
