siRNA Details
Gene Symbol: Grifin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030022.1 | NP_084298.1 | 4 | 304 | ||
GenBank mRNA | AK021365.1 | - | 4 | 305 | ||
BC107323.2 | - | 4 | 304 | 40053455 | ||
BC107324.1 | - | 4 | 304 | 40053456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
