siRNA Details
Gene Symbol: Tmem190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030028.1 | NP_084304.1 | 5 | 381 | ||
GenBank mRNA | AK019807.1 | - | 5 | 374 | ||
BU936271.1 | - | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
