siRNA Details
Gene Symbol: Cpt1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252470.1 | NP_001239399.1 | 17 | 2361 | ||
NM_153679.2 | NP_710146.1 | 17 | 2363 | |||
GenBank mRNA | AF320000.1 | - | 17 | 2286 | ||
AK035790.1 | - | 17 | 2327 | |||
AK087747.1 | - | 17 | 2257 | |||
BC043460.1 | - | 8 | 1410 | |||
BC066155.1 | - | 17 | 2285 | 30089333 | ||
BC083317.1 | - | 17 | 2244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
