siRNA Details
Gene Symbol: Sowahb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175270.4 | NP_780479.1 | 1 | 2196 | ||
GenBank mRNA | AK033423.1 | - | 1 | 2126 | ||
AK033641.1 | - | 1 | 1659 | |||
AK137429.1 | - | 1 | 2197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
