siRNA Details
Gene Symbol: Trappc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020026.1 | - | 3 | 368 | ||
AK046610.1 | - | 3 | 396 | |||
AK089083.1 | - | 3 | 209 | |||
AK168294.1 | - | 4 | 656 | |||
BC031464.1 | - | 3 | 353 | 4017482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
