siRNA Details
Gene Symbol: Cst11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030059.2 | NP_084335.1 | 1 | 258 | ||
GenBank mRNA | AF454373.1 | - | 1 | 260 | ||
AK020300.1 | - | 1 | 260 | |||
BC100526.1 | - | 1 | 183 | 30920256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
