siRNA Details
Gene Symbol: Acbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030063.2 | NP_084339.1 | 3 | 184 | ||
GenBank mRNA | AK020348.1 | - | 3 | 185 | ||
AV497738.1 | - | 3 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
