siRNA Details
Gene Symbol: Zfp687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030074.2 | NP_084350.1 | 9 | 3484 | ||
GenBank mRNA | AK019851.2 | - | 9 | 3485 | ||
AK129360.1 | - | 3 | 3670 | |||
AK133173.1 | - | 6 | 4193 | |||
AK148239.1 | - | 8 | 3599 | |||
BC034652.1 | - | 3 | 317 | |||
BC036979.1 | - | 4 | 723 | |||
BC039950.1 | - | 4 | 697 | |||
BC056943.1 | - | 7 | 3134 | 6831344 | ||
BC060179.1 | - | 7 | 3134 | 6831344 |

More Information
Other Species Targeted

Gene Ontology Categories:
